Learn More
Thermo Scientific Chemicals N-Acetyl-L-glutamic acid, 99%
CAS: 1188-37-0 | C7H11NO5 | 189.167 g/mol
Supplier: Thermo Scientific Chemicals B2362122
Description
N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.
Solubility
Soluble in water. 2.7 g/100 mL (20°C)
Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
| 194°C to 199°C | |
| 1188-37-0 | |
| MFCD00002802 | |
| n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh | |
| RFMMMVDNIPUKGG-YFKPBYRVSA-N | |
| (2S)-2-acetamidopentanedioic acid | |
| 189.167 | |
| CHEBI:17533 | |
| 189.17 |
| N-Acetyl-L-glutamic acid | |
| C7H11NO5 | |
| 1727473 | |
| Soluble in water. 2.7g/100 mL (20°C) | |
| CC(=O)NC(CCC(=O)O)C(=O)O | |
| 100 g | |
| 70914 | |
| −16.5° (c=1 in Water) | |
| 99% |
Chemical Identifiers
| 1188-37-0 | |
| 189.167 | |
| RFMMMVDNIPUKGG-YFKPBYRVSA-N | |
| 70914 | |
| (2S)-2-acetamidopentanedioic acid |
| C7H11NO5 | |
| MFCD00002802 | |
| n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh | |
| CHEBI:17533 | |
| CC(=O)NC(CCC(=O)O)C(=O)O |
Safety and Handling
EINECSNumber : 214-708-4
RTECSNumber : LZ9725000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only