missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-glutamic acid, 99%

Catalog No. AAB2362114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2362114 25 g
AAB2362122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2362114 Supplier Thermo Scientific Chemicals Supplier No. B2362114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1188-37-0 | C7H11NO5 | 189.167 g/mol

N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water. 2.7 g/100 mL (20°C)

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Chemical Identifiers

CAS 1188-37-0
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.167
MDL Number MFCD00002802
InChI Key RFMMMVDNIPUKGG-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
PubChem CID 70914
ChEBI CHEBI:17533
IUPAC Name (2S)-2-acetamidopentanedioic acid
SMILES CC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 194°C to 199°C
Beilstein 1727473
Solubility Information Soluble in water. 2.7g/100 mL (20°C)
Quantity 25 g
Formula Weight 189.17
Optical Rotation −16.5° (c=1 in Water)
Percent Purity 99%
Chemical Name or Material N-Acetyl-L-glutamic acid
EINECSNumber 214-708-4
RTECSNumber LZ9725000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only