missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-cysteine, 98%

Catalog No. AC160282500
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160282500 250 g Glass bottle
AC160280250 25 g Glass bottle
AC160280010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160282500 Supplier Thermo Scientific Chemicals Supplier No. 160282500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 616-91-1 | C5H9NO3S | 163.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-91-1
Molecular Formula C5H9NO3S
Molecular Weight (g/mol) 163.19
MDL Number MFCD00004880
InChI Key PWKSKIMOESPYIA-BYPYZUCNSA-N
Synonym n-acetyl-l-cysteine, acetylcysteine, n-acetylcysteine, mercapturic acid, acetadote, l-acetylcysteine, broncholysin, fluimucil, mucomyst, fluprowit
PubChem CID 12035
ChEBI CHEBI:28939
IUPAC Name (2R)-2-acetamido-3-sulfanylpropanoic acid
SMILES CC(=O)NC(CS)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 109.0°C to 111.0°C
Color White
Ignition Residue 0.5% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HSCH2CH(NHCOCH3)CO2H
Quantity 250 g
Merck Index 15, 83
Specific Rotation Condition +5.50° (20°C c=2,H2O)
Specific Rotation + 5.50
Solubility Information Solubility in water: soluble in water.
Formula Weight 163.19
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-Acetyl-L-cysteine
Show More Show Less
EINECSNumber 210-498-3

RUO – Research Use Only