missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-DL-serine, 98+%, Thermo Scientific Chemicals
$774.97 - $774.97
Chemical Identifiers
| CAS | 97-14-3 |
|---|---|
| Molecular Formula | C5H9NO4 |
| Molecular Weight (g/mol) | 147.13 |
| MDL Number | MFCD00037248 |
| InChI Key | JJIHLJJYMXLCOY-UHFFFAOYSA-N |
| Synonym | n-acetyl-dl-serine, 2-acetylamino-3-hydroxypropanoic acid, 2-acetylamino-3-hydroxypropionic acid, l-serine, n-acetyl, 2-acetamido-3-oxidanyl-propanoic acid, n-acetylserine #, dl-serine, n-acetyl, serine, n-acetyl-, l |
| PubChem CID | 352294 |
| IUPAC Name | 2-acetamido-3-hydroxypropanoic acid |
| SMILES | CC(=O)NC(CO)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2771706
|
Thermo Scientific Chemicals
H2771706 |
5 g |
Each for $774.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 97-14-3 | |
| 147.13 | |
| JJIHLJJYMXLCOY-UHFFFAOYSA-N | |
| 352294 | |
| CC(=O)NC(CO)C(=O)O |
| C5H9NO4 | |
| MFCD00037248 | |
| n-acetyl-dl-serine, 2-acetylamino-3-hydroxypropanoic acid, 2-acetylamino-3-hydroxypropionic acid, l-serine, n-acetyl, 2-acetamido-3-oxidanyl-propanoic acid, n-acetylserine #, dl-serine, n-acetyl, serine, n-acetyl-, l | |
| 2-acetamido-3-hydroxypropanoic acid |
Specifications
| 97-14-3 | |
| MFCD00037248 | |
| JJIHLJJYMXLCOY-UHFFFAOYSA-N | |
| 2-acetamido-3-hydroxypropanoic acid | |
| 147.13 | |
| 147.13 | |
| N-Acetyl-DL-serine |
| C5H9NO4 | |
| n-acetyl-dl-serine, 2-acetylamino-3-hydroxypropanoic acid, 2-acetylamino-3-hydroxypropionic acid, l-serine, n-acetyl, 2-acetamido-3-oxidanyl-propanoic acid, n-acetylserine #, dl-serine, n-acetyl, serine, n-acetyl-, l | |
| CC(=O)NC(CO)C(=O)O | |
| 5 g | |
| 352294 | |
| ≥98% |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only