missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-serine, 98+%, Thermo Scientific Chemicals

Catalog No. AAH2771706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2771706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2771706 Supplier Thermo Scientific Chemicals Supplier No. H2771706
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-14-3
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00037248
InChI Key JJIHLJJYMXLCOY-UHFFFAOYSA-N
Synonym n-acetyl-dl-serine, 2-acetylamino-3-hydroxypropanoic acid, 2-acetylamino-3-hydroxypropionic acid, l-serine, n-acetyl, 2-acetamido-3-oxidanyl-propanoic acid, n-acetylserine #, dl-serine, n-acetyl, serine, n-acetyl-, l
PubChem CID 352294
IUPAC Name 2-acetamido-3-hydroxypropanoic acid
SMILES CC(=O)NC(CO)C(=O)O

Specifications

Quantity 5 g
Formula Weight 147.13
Percent Purity ≥98%
Chemical Name or Material N-Acetyl-DL-serine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only