missing translation for 'onlineSavingsMsg'
Learn More

N-acétyl-DL-phénylalanine 98,0+%, TCI America™

Numéro de catalogue. p-7118118
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A28645G 5 g
A286425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A28645G Fournisseur TCI America Code fournisseur A28645G

Identifiants chimiques

CAS 2901-75-9
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD00063157
InChI Key CBQJSKKFNMDLON-UHFFFAOYSA-N
Synonym n-acetyl-dl-phenylalanine, afalanine, n-acetylphenylalanine, ac-dl-phe-oh, n-acetyl-d,l-phenylalanine, afalanina, afalaninum, dl-n-acetyl-3-phenylalanine, afalanine inn, dl-2-acetamido-3-phenylpropionic acid
PubChem CID 2000
ChEBI CHEBI:21626
IUPAC Name 2-acétamido-3-phénylpropanoïque
SMILES CC(=O)NC(CC1=CC=CC=C1)C(=O)O

Spécifications

Melting Point 154°C
Color Blanc
Quantity 5 g
Formula Weight 207.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Poudre cristalline
Chemical Name or Material N-Acetyl-DL-phenylalanine
EINECSNumber (9)-1617
RTECSNumber SQ7321132
TSCA No