missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-methionine 98.0+%, TCI America™

Catalog No. p-7116715
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A010025G 25 g
A0100500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A010025G Supplier TCI America Supplier No. A010025G
Only null left

Chemical Identifiers

CAS 1115-47-5
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00008681
InChI Key XUYPXLNMDZIRQH-UHFFFAOYNA-N
Synonym n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
PubChem CID 6180
IUPAC Name 2-acetamido-4-(methylsulfanyl)butanoic acid
SMILES CSCCC(NC(C)=O)C(O)=O

Specifications

Melting Point 116°C
Color White
Quantity 25 g
Formula Weight 191.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-DL-methionine
EINECSNumber (9)-1631
RTECSNumber PD0500000
TSCA Yes