missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99%

CAS: 1115-47-5 | C7H13NO3S | 191.25 g/mol

$38.82 - $179.32

Chemical Identifiers

CAS 1115-47-5
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00008681
InChI Key XUYPXLNMDZIRQH-UHFFFAOYNA-N
Synonym n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
PubChem CID 6180
SMILES CSCCC(NC(C)=O)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2186618
View Documents
Thermo Scientific Chemicals
B2186618
50 g
Each for $38.82
Only null left
 
AAB2186630
View Documents
Thermo Scientific Chemicals
B2186630
250 g
Each for $71.73
Only null left
 
AAB218660B
View Documents
Thermo Scientific Chemicals
B218660B
1000 g
Each for $179.32
Only null left
 
Description

Description

N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

Solubility
Soluble in water, ethanol, ethyl acetate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

1115-47-5
191.25
XUYPXLNMDZIRQH-UHFFFAOYNA-N
6180
C7H13NO3S
MFCD00008681
n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
CSCCC(NC(C)=O)C(O)=O

Specifications

113°C to 117°C
C7H13NO3S
1725554
n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
XUYPXLNMDZIRQH-UHFFFAOYNA-N
2-acetamido-4-(methylsulfanyl)butanoic acid
191.25
191.25
N-Acetyl-DL-methionine
1115-47-5
MFCD00008681
14,96
Soluble in water,ethanol,ethyl acetate.
CSCCC(NC(C)=O)C(O)=O
50 g
6180
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-224-3

RTECSNumber : PD0500000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only