missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99%

Catalog No. AAB2186618
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2186618 50 g
AAB2186630 250 g
AAB218660B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2186618 Supplier Thermo Scientific Chemicals Supplier No. B2186618

CAS: 1115-47-5 | C7H13NO3S | 191.25 g/mol

N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

Solubility
Soluble in water, ethanol, ethyl acetate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1115-47-5
Molecular Formula C7H13NO3S
Molecular Weight (g/mol) 191.25
MDL Number MFCD00008681
InChI Key XUYPXLNMDZIRQH-UHFFFAOYNA-N
Synonym n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
PubChem CID 6180
SMILES CSCCC(NC(C)=O)C(O)=O

Specifications

Melting Point 113°C to 117°C
Beilstein 1725554
Merck Index 14,96
Solubility Information Soluble in water,ethanol,ethyl acetate.
IUPAC Name 2-acetamido-4-(methylsulfanyl)butanoic acid
Quantity 50 g
Formula Weight 191.25
Percent Purity 99%
Chemical Name or Material N-Acetyl-DL-methionine
EINECSNumber 214-224-3
RTECSNumber PD0500000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only