missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-DL-leucine 99.0+%, TCI America™
$69.31 - $111.66
Chemical Identifiers
| CAS | 99-15-0 |
|---|---|
| Molecular Formula | C8H15NO3 |
| Molecular Weight (g/mol) | 173.21 |
| MDL Number | MFCD00026498 |
| InChI Key | WXNXCEHXYPACJF-UHFFFAOYNA-N |
| Synonym | n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid |
| PubChem CID | 1995 |
| IUPAC Name | 2-acetamido-4-methylpentanoic acid |
| SMILES | CC(C)CC(NC(C)=O)C(O)=O |
Chemical Identifiers
| 99-15-0 | |
| 173.21 | |
| WXNXCEHXYPACJF-UHFFFAOYNA-N | |
| 1995 | |
| CC(C)CC(NC(C)=O)C(O)=O |
| C8H15NO3 | |
| MFCD00026498 | |
| n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid | |
| 2-acetamido-4-methylpentanoic acid |
Specifications
| 99-15-0 | |
| White | |
| MFCD00026498 | |
| n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid | |
| CC(C)CC(NC(C)=O)C(O)=O | |
| 173.21 | |
| 173.21 | |
| Crystalline Powder |
| 160°C | |
| C8H15NO3 | |
| 5 g | |
| WXNXCEHXYPACJF-UHFFFAOYNA-N | |
| 2-acetamido-4-methylpentanoic acid | |
| 1995 | |
| ≥99.0% (T) | |
| N-Acetyl-DL-leucine |
Safety and Handling
TSCA : No