missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-leucine, 99%

CAS: 99-15-0 | C8H15NO3 | 173.21 g/mol

Supplier:  Thermo Scientific Chemicals H2744114

 View more versions of this product

Catalog No. AAH2744114

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

159°C to 161°C
99-15-0
MFCD00026498
WXNXCEHXYPACJF-UHFFFAOYNA-N
2-acetamido-4-methylpentanoic acid
173.21
173.21
N-Acetyl-DL-leucine
C8H15NO3
n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid
CC(C)CC(NC(C)=O)C(O)=O
25 g
1995
99%

Chemical Identifiers

99-15-0
173.21
WXNXCEHXYPACJF-UHFFFAOYNA-N
1995
CC(C)CC(NC(C)=O)C(O)=O
C8H15NO3
MFCD00026498
n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid
2-acetamido-4-methylpentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 202-734-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only