missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-DL-leucine, 99%

Catalog No. AAH2744106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2744106 5 g
AAH2744114 25 g
AAH2744122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2744106 Supplier Thermo Scientific Chemicals Supplier No. H2744106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-15-0 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-15-0
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00026498
InChI Key WXNXCEHXYPACJF-UHFFFAOYNA-N
Synonym n-acetyl-dl-leucine, acetylleucine, ac-dl-leu-oh, tanganil, l-leucine, n-acetyl, acetyl-dl-leucine, acetylleucine inn, dl-leucine, n-acetyl, 2-acetylamino-4-methylpentanoic acid
PubChem CID 1995
IUPAC Name 2-acetamido-4-methylpentanoic acid
SMILES CC(C)CC(NC(C)=O)C(O)=O

Specifications

Melting Point 159°C to 161°C
Quantity 5 g
Formula Weight 173.21
Percent Purity 99%
Chemical Name or Material N-Acetyl-DL-leucine
EINECSNumber 202-734-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only