missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-DL-histidine Monohydrate 98.0+%, TCI America™
$85.75 - $257.24
Chemical Identifiers
| CAS | 213178-97-3 |
|---|---|
| Molecular Formula | C8H13N3O4 |
| Molecular Weight (g/mol) | 215.209 |
| MDL Number | MFCD00065961 |
| InChI Key | PSWSDQRXCOJSFC-UHFFFAOYSA-N |
| Synonym | 2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, ac-dl-his-oh.h2o, n-acetyl-dl-histidine monohydrate, 2-acetamido-3-1h-imidazol-5-yl propanoic acid hydrate, 2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, n-acetyl-l-histidine monohydrate, 2-acetamido-3-3h-imidazol-4-yl propanoic acid hydrate, acmc-1c6iz, acmc-209j3m, n-acetyl-dl-histidine hydrate |
| PubChem CID | 18668507 |
| IUPAC Name | 2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate |
| SMILES | CC(=O)NC(CC1=CN=CN1)C(=O)O.O |
Chemical Identifiers
| 213178-97-3 | |
| 215.209 | |
| PSWSDQRXCOJSFC-UHFFFAOYSA-N | |
| 18668507 | |
| CC(=O)NC(CC1=CN=CN1)C(=O)O.O |
| C8H13N3O4 | |
| MFCD00065961 | |
| 2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, ac-dl-his-oh.h2o, n-acetyl-dl-histidine monohydrate, 2-acetamido-3-1h-imidazol-5-yl propanoic acid hydrate, 2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, n-acetyl-l-histidine monohydrate, 2-acetamido-3-3h-imidazol-4-yl propanoic acid hydrate, acmc-1c6iz, acmc-209j3m, n-acetyl-dl-histidine hydrate | |
| 2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate |
Specifications
| 213178-97-3 | |
| White | |
| MFCD00065961 | |
| 2-acetamido-3-1h-imidazol-4-yl propanoic acid hydrate, ac-dl-his-oh.h2o, n-acetyl-dl-histidine monohydrate, 2-acetamido-3-1h-imidazol-5-yl propanoic acid hydrate, 2-acetylamino-3-1h-imidazol-4-yl propanoic acid hydrate, n-acetyl-l-histidine monohydrate, 2-acetamido-3-3h-imidazol-4-yl propanoic acid hydrate, acmc-1c6iz, acmc-209j3m, n-acetyl-dl-histidine hydrate | |
| CC(=O)NC(CC1=CN=CN1)C(=O)O.O | |
| 215.209 | |
| 197.19 | |
| Crystalline Powder |
| 152°C | |
| C8H13N3O4 | |
| 1 g | |
| PSWSDQRXCOJSFC-UHFFFAOYSA-N | |
| 2-acetamido-3-(1H-imidazol-5-yl)propanoic acid;hydrate | |
| 18668507 | |
| ≥98.0% (HPLC,T) | |
| N-Acetyl-DL-histidine Monohydrate |
Safety and Handling
TSCA : No