missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-alanine, 97+%

Catalog No. AAL1032906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1032906 5 g
AAL1032914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1032906 Supplier Thermo Scientific Chemicals Supplier No. L1032906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1115-69-1 | C5H9NO3 | 131.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1115-69-1
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.131
MDL Number MFCD00037238
InChI Key KTHDTJVBEPMMGL-UHFFFAOYSA-N
Synonym n-acetyl-dl-alanine, ac-dl-ala-oh, 2-acetylaminopropionic acid, n-acetylalanine, acetyl-dl-alanine, dl-alanine, n-acetyl, 2-acetylamino-propionic acid, n-acetyl-s-alanine, n-alpha-acetyl-dl-alanine, n-acetyl-dl-2-aminopropionic acid
PubChem CID 7345
IUPAC Name 2-acetamidopropanoic acid
SMILES CC(C(=O)O)NC(=O)C

Specifications

Melting Point 135°C to 139°C
Beilstein 774333
Quantity 5 g
Formula Weight 131.13
Percent Purity ≥97%
Chemical Name or Material N-Acetyl-DL-alanine
EINECSNumber 214-229-0
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only