missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-mannosamine monohydrate, 99%

Catalog No. AAL1116703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1116703 1 g
AAL1116706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1116703 Supplier Thermo Scientific Chemicals Supplier No. L1116703

CAS: 1071625-31-4 | C8H17NO7 | 239.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1071625-31-4
Molecular Formula C8H17NO7
Molecular Weight (g/mol) 239.22
MDL Number MFCD00149493,MFCD00046758,MFCD00149493,MFCD00069808
InChI Key VVQPUTSNIMAJPT-UHFFFAOYNA-N
Synonym n-acetylmannosamine
PubChem CID 11908605
IUPAC Name N-[(2R,3S,4S,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES O.CC(=O)NC1C(O)OC(CO)C(O)C1O

Specifications

Melting Point ∼130°C (decomposition)
Packaging Glass bottle
Quantity 1 g
Beilstein 1346524
Formula Weight 239.24 (221.22 anhydrous)
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-D-mannosamine monohydrate
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only