missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-glucosamine 98.0+%, TCI America™
$68.97 - $730.71
Chemical Identifiers
| CAS | 7512-17-6 |
|---|---|
| Molecular Formula | C8H15NO6 |
| Molecular Weight (g/mol) | 221.21 |
| MDL Number | MFCD00061615 |
| InChI Key | OVRNDRQMDRJTHS-WZWLWIOANA-N |
| Synonym | 2-Acetamido-2-deoxy-D-glucopyranose |
| IUPAC Name | N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
| SMILES | CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
Chemical Identifiers
| 7512-17-6 | |
| 221.21 | |
| OVRNDRQMDRJTHS-WZWLWIOANA-N | |
| N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
| C8H15NO6 | |
| MFCD00061615 | |
| 2-Acetamido-2-deoxy-D-glucopyranose | |
| CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
Specifications
| 7512-17-6 | |
| White | |
| MFCD00061615 | |
| 2-Acetamido-2-deoxy-D-glucopyranose | |
| CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O | |
| 221.21 | |
| ≥98.0% (HPLC,N) | |
| N-Acetyl-D-glucosamine |
| 205°C | |
| C8H15NO6 | |
| 25 g | |
| OVRNDRQMDRJTHS-WZWLWIOANA-N | |
| N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide | |
| 221.21 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (8)-0097
TSCA : Yes