missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-D-glucosamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 205°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 221.21 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Acetyl-D-glucosamine |
Safety and Handling
| EINECSNumber | (8)-0097 |
| TSCA | Yes |