missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-glucosamine, 98+%

Catalog No. AAA1304718
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1304718 50 g
AAA1304709 10 g
AAA1304722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1304718 Supplier Thermo Scientific Chemicals Supplier No. A1304718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-Acetyl-D-glucosamine, CAS # 72-87-7, is the D isomer of the glucose-derived monomer found in structural polysaccharides.

N-Acetyl-D-glucosamine is used in the identification and characterization of N-acetyl-beta-D-hexoaminidase. It is also used in the treatment of osteoarthritis and inflammatory bowel disease (IBD), including ulcerative colitis and Crohn's disease. Further, it is used as a substrate in sialic acid production, cosmetics and in drug development research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• N-Acetyl-D-glucosamine is the acetylated form of glucosamine, a glucose derivative

• It has a role as a prokaryotic metabolism and monomer used in chitin, hyaluronic acid, and polymers within the cell wall

Application

• N-Acetyl-D-Glucosamine can be used for molecular binding studies; research has shown that it attaches to certain microbes and their pathogens

• It is useful for biochemical characterization studies of related glucosamines

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Melting Point 213°C (decomposition)
Assay Percent Range ≥98%
Quantity 50 g
Beilstein 1727589
Sensitivity Hygroscopic
Solubility Information Soluble in concentrated hydrochloric acid,sulfuric acid,phosphoric acid and formic acid. Insoluble in water,dilute acids,dilute alkalis,concentrated alkalis and organic solvents.
Formula Weight 221.21
Chemical Name or Material N-Acetyl-D-glucosamine
EINECSNumber 231-368-2
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only