missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-D-glucosamine, 100%, MP Biomedicals™

Catalog No. ICN10006880
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10006880 100 g
ICN10006825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10006880 Supplier MP Biomedicals Inc Supplier No. 0210006880

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7512-17-6
Molecular Formula C8H15NO6
Molecular Weight (g/mol) 221.21
MDL Number MFCD00061615
InChI Key OVRNDRQMDRJTHS-WZWLWIOANA-N
Synonym 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc
IUPAC Name N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
SMILES CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O

Specifications

Color White
Assay Percent Range 1
Quantity 100 g
Formula Weight 221.2
Grade Not Available
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Acetyl-D-glucosamine