missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-beta-alanine, 97%

Catalog No. AAH5020806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5020806 5 g
AAH5020803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5020806 Supplier Thermo Scientific Chemicals Supplier No. H5020806

CAS: 3025-95-4 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3025-95-4
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00037296
InChI Key LJLLAWRMBZNPMO-UHFFFAOYSA-N
Synonym n-acetyl-beta-alanine, n-acetyl-b-alanine, ac-beta-ala-oh, n-acetyl-, a-alanine, 3-acetylamino propanoic acid, n-acetyl-?-alanine, acetyl-beta-alanine, n-acetyl-ss-alanine, ac--ala-oh, beta-alanine, n-acetyl
PubChem CID 76406
ChEBI CHEBI:16682
IUPAC Name 3-acetamidopropanoic acid
SMILES CC(=O)NCCC(O)=O

Specifications

Melting Point 77°C
Linear Formula CH3CONHCH2CH2CO2H
Quantity 5 g
Formula Weight 131.13
Percent Purity 97%
Chemical Name or Material N-Acetyl-beta-alanine
EINECSNumber 221-185-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only