missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-Asp-Glu-Val-Asp-7-amino-4-(trifluoromethyl)coumarin

Catalog No. AAJ64134LB
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64134LB 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64134LB Supplier Thermo Scientific Chemicals Supplier No. J64134LB0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

A fluorogenic substrate for caspase-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 201608-14-2
Molecular Formula C30H34F3N5O13
Synonym Ac-DEVD-AFC

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A Fluorogenic Substrate for Caspase-3
Formula Weight 729.61 g/mol
Product Type Signal Transduction Reagent
Form Powder

RUO – Research Use Only