missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-5-hydroxytryptamine 98.0+%, TCI America™
$140.54 - $658.41
Chemical Identifiers
| CAS | 1210-83-9 |
|---|---|
| Molecular Formula | C12H14N2O2 |
| Molecular Weight (g/mol) | 218.26 |
| MDL Number | MFCD00005656 |
| InChI Key | MVAWJSIDNICKHF-UHFFFAOYSA-N |
| Synonym | n-acetyl-5-hydroxytryptamine, n-acetylserotonin, n-acetyl serotonin, normelatonin, n-2-5-hydroxy-1h-indol-3-yl ethyl acetamide, o-demethylmelatonin, 5-hydroxy-n-acetyltryptamine, acetylserotonin, unii-p4to3c82wv, 5-hydroxymelatonin |
| PubChem CID | 903 |
| ChEBI | CHEBI:17697 |
| IUPAC Name | N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide |
| SMILES | CC(=O)NCCC1=CNC2=CC=C(O)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1277100MG
|
TCI America
A1277100MG |
100 mg |
Each for $140.54
|
|
|||||
|
A12771G
|
TCI America
A12771G |
1 g |
Each for $658.41
|
|
|||||
Chemical Identifiers
| 1210-83-9 | |
| 218.26 | |
| MVAWJSIDNICKHF-UHFFFAOYSA-N | |
| 903 | |
| N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide |
| C12H14N2O2 | |
| MFCD00005656 | |
| n-acetyl-5-hydroxytryptamine, n-acetylserotonin, n-acetyl serotonin, normelatonin, n-2-5-hydroxy-1h-indol-3-yl ethyl acetamide, o-demethylmelatonin, 5-hydroxy-n-acetyltryptamine, acetylserotonin, unii-p4to3c82wv, 5-hydroxymelatonin | |
| CHEBI:17697 | |
| CC(=O)NCCC1=CNC2=CC=C(O)C=C12 |
Specifications
| 1210-83-9 | |
| White-Yellow | |
| MFCD00005656 | |
| n-acetyl-5-hydroxytryptamine, n-acetylserotonin, n-acetyl serotonin, normelatonin, n-2-5-hydroxy-1h-indol-3-yl ethyl acetamide, o-demethylmelatonin, 5-hydroxy-n-acetyltryptamine, acetylserotonin, unii-p4to3c82wv, 5-hydroxymelatonin | |
| CC(=O)NCCC1=CNC2=CC=C(O)C=C12 | |
| 218.26 | |
| CHEBI:17697 | |
| ≥98.0% (HPLC,N) | |
| N-Acetyl-5-hydroxytryptamine |
| 120°C | |
| C12H14N2O2 | |
| 100 mg | |
| MVAWJSIDNICKHF-UHFFFAOYSA-N | |
| N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]acetamide | |
| 903 | |
| 218.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No