Learn More
N-Acetonylpyridinium chloride, 97%
CAS: 42508-60-1 | C8H10ClNO | 171.62 g/mol
$105.21 - $367.18
Chemical Identifiers
| CAS | 42508-60-1 |
|---|---|
| Molecular Formula | C8H10ClNO |
| Molecular Weight (g/mol) | 171.62 |
| MDL Number | MFCD00031949 |
| InChI Key | JHYYWKUENNZTMD-UHFFFAOYSA-M |
| Synonym | 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride |
| PubChem CID | 2734121 |
| SMILES | [Cl-].CC(=O)C[N+]1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1601506
|
Thermo Scientific Chemicals
A1601506 |
5 g |
Each for $105.21
|
|
|||||
|
AAA1601514
|
Thermo Scientific Chemicals
A1601514 |
25 g |
Each for $367.18
|
|
|||||
Description
N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from water and humidity.
Chemical Identifiers
| 42508-60-1 | |
| 171.62 | |
| JHYYWKUENNZTMD-UHFFFAOYSA-M | |
| 2734121 |
| C8H10ClNO | |
| MFCD00031949 | |
| 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride | |
| [Cl-].CC(=O)C[N+]1=CC=CC=C1 |
Specifications
| 42508-60-1 | |
| MFCD00031949 | |
| Hygroscopic | |
| Soluble in water. | |
| [Cl-].CC(=O)C[N+]1=CC=CC=C1 | |
| 171.62 | |
| 171.63 | |
| N-Acetonylpyridinium chloride |
| C8H10ClNO | |
| 5 g | |
| 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride | |
| JHYYWKUENNZTMD-UHFFFAOYSA-M | |
| 1-(2-oxopropyl)pyridin-1-ium chloride | |
| 2734121 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only