missing translation for 'onlineSavingsMsg'
Learn More

N-Acetonylpyridinium chloride, 97%

Catalog No. AAA1601506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1601506 5 g
AAA1601514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1601506 Supplier Thermo Scientific Chemicals Supplier No. A1601506

CAS: 42508-60-1 | C8H10ClNO | 171.62 g/mol

N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from water and humidity.

Chemical Identifiers

CAS 42508-60-1
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00031949
InChI Key JHYYWKUENNZTMD-UHFFFAOYSA-M
Synonym 1-acetonylpyridinium chloride, n-acetonylpyridinium chloride, 1-2-oxopropyl pyridinium chloride, 1-2-oxopropyl pyridin-1-ium chloride, acmc-209jpw, 1-acetonylpyridiniumchloride, 1-pyridin-1-ium-1-ylpropan-2-one chloride, 1-2-oxopropyl-2h-pyridin-2-ylium chloride
PubChem CID 2734121
SMILES [Cl-].CC(=O)C[N+]1=CC=CC=C1

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
IUPAC Name 1-(2-oxopropyl)pyridin-1-ium chloride
Formula Weight 171.63
Percent Purity 97%
Chemical Name or Material N-Acetonylpyridinium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only