missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine 98.0+%, TCI America™

Catalog No. F10281G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10281G Supplier TCI America Supplier No. F10281G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103478-62-2
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00151933
InChI Key BUJQSIPFDWLNDC-GNLPSFAGNA-N
Synonym fmoc-n-methyl-l-leucine, fmoc-n-me-leu-oh, n-fmoc-n-methyl-l-leucine, fmoc-meleu-oh, l-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, pubchem13155, fmoc-n-a-methyl-l-leucine, n-methyl-n-fmoc-l-leu-oh
PubChem CID 7015835
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-4-methylpentanoic acid
SMILES CC(C)C[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 1 g
Formula Weight 367.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-leucine