missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-isoleucine 98.0+%, TCI America™

Catalog No. p-7130076
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F11291G 1 g
F11295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F11291G Supplier TCI America Supplier No. F11291G
Only null left

Chemical Identifiers

CAS 138775-22-1
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00153389
InChI Key IQIOLCJHRZWOLS-DZLYFNQKNA-N
Synonym fmoc-n-me-ile-oh, fmoc-n-methyl-l-isoleucine, fmoc-n-me-lle-oh, n-fmoc-n-methyl-l-isoleucine, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-isoleucine, 2s,3s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylpentanoic acid, fmoc-melle-oh, fmoc-l-meile-oh, ambotzfaa1398, pubchem18966
PubChem CID 15433034
IUPAC Name (2S,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 1 g
Formula Weight 367.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-isoleucine
TSCA No
Recommended Storage Refrigerator