missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine Hydrate 97.0+%, TCI America™

Catalog No. F04541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04541G 1 g
F04545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04541G Supplier TCI America Supplier No. F04541G
Only null left

Chemical Identifiers

CAS 73724-45-5
Molecular Formula C18H16NO5
Molecular Weight (g/mol) 326.33
MDL Number MFCD00051928
InChI Key JZTKZVJMSCONAK-INIZCTEOSA-M
Synonym fmoc-ser-oh, fmoc-l-serine, fmoc-l-ser-oh, n-fmoc-l-serine, n-9-fluorenylmethoxycarbonyl-l-serine, n-9h-fluoren-9-ylmethoxy carbonyl-l-serine, n-9h-fluoren-9-ylmethoxycarbonyl serine, s-fmoc-2-amino-3-hydroxypropionic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-serine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid
PubChem CID 6541433
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxypropanoate
SMILES OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 327.34
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine Hydrate