missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 98.0+%, TCI America™
$37.29 - $59.65
Chemical Identifiers
| CAS | 121343-82-6 |
|---|---|
| Molecular Formula | C20H19NO6 |
| Molecular Weight (g/mol) | 369.373 |
| MDL Number | MFCD00237657 |
| InChI Key | QEPWHIXHJNNGLU-KRWDZBQOSA-N |
| Synonym | fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid |
| PubChem CID | 7019018 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O |
Chemical Identifiers
| 121343-82-6 | |
| 369.373 | |
| QEPWHIXHJNNGLU-KRWDZBQOSA-N | |
| 7019018 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O |
| C20H19NO6 | |
| MFCD00237657 | |
| fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid |
Specifications
| 121343-82-6 | |
| C20H19NO6 | |
| 1 g | |
| QEPWHIXHJNNGLU-KRWDZBQOSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid | |
| 7019018 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid |
| White | |
| MFCD00237657 | |
| fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O | |
| 369.373 | |
| 369.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : No