missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 98.0+%, TCI America™

Catalog No. F04535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04535G 5 g
F04531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04535G Supplier TCI America Supplier No. F04535G

Chemical Identifiers

CAS 121343-82-6
Molecular Formula C20H19NO6
Molecular Weight (g/mol) 369.373
MDL Number MFCD00237657
InChI Key QEPWHIXHJNNGLU-KRWDZBQOSA-N
Synonym fmoc-glu-oh, fmoc-l-glutamic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pentanedioic acid, n-fmoc-l-glutamic acid, fmoc-glu, n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamic acid, l-glutamic acid, n-9h-fluoren-9-ylmethoxy carbonyl, 2s-2-fluoren-9-ylmethoxy carbonylamino pentanedioic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanedioic acid
PubChem CID 7019018
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 369.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid