missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 369.37 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic Acid |
Safety and Handling
| TSCA | No |