missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 98.0+%, TCI America™
$37.29 - $262.83
Chemical Identifiers
| CAS | 119062-05-4 |
|---|---|
| Molecular Formula | C19H17NO6 |
| Molecular Weight (g/mol) | 355.346 |
| MDL Number | MFCD00237654 |
| InChI Key | KSDTXRUIZMTBNV-INIZCTEOSA-N |
| Synonym | fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid |
| PubChem CID | 7019016 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O |
Chemical Identifiers
| 119062-05-4 | |
| 355.346 | |
| KSDTXRUIZMTBNV-INIZCTEOSA-N | |
| 7019016 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O |
| C19H17NO6 | |
| MFCD00237654 | |
| fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid |
Specifications
| 119062-05-4 | |
| C19H17NO6 | |
| 1 g | |
| KSDTXRUIZMTBNV-INIZCTEOSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid | |
| 7019016 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid |
| White | |
| MFCD00237654 | |
| fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O | |
| 355.346 | |
| 355.35 | |
| Crystalline Powder |
Safety and Handling
TSCA : No