missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 98.0+%, TCI America™

Catalog No. F04521G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04521G 1 g
F04525G 5 g
F045225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F04521G Supplier TCI America Supplier No. F04521G

Chemical Identifiers

CAS 119062-05-4
Molecular Formula C19H17NO6
Molecular Weight (g/mol) 355.346
MDL Number MFCD00237654
InChI Key KSDTXRUIZMTBNV-INIZCTEOSA-N
Synonym fmoc-asp-oh, fmoc-l-aspartic acid, n-fmoc-l-aspartic acid, fmoc-l-asp-oh, fmoc-aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino succinic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino succinic acid, aspartic acid, n-9h-fluoren-9-ylmethoxy carbonyl, n-9-fluorenylmethoxycarbonyl-l-aspartic acid
PubChem CID 7019016
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanedioic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 355.35
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid