missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine 98.0+%, TCI America™

Catalog No. p-7129779
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06055G 5 g
F060525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06055G Supplier TCI America Supplier No. F06055G

Chemical Identifiers

CAS 86123-10-6
Molecular Formula C24H21NO4
Molecular Weight (g/mol) 387.44
MDL Number MFCD00062955
InChI Key SJVFAHZPLIXNDH-JOCHJYFZSA-N
Synonym fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
PubChem CID 978332
IUPAC Name (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-phenylpropanoic acid
SMILES OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 183°C
Color White
Quantity 5 g
Formula Weight 387.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine