missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine 98.0+%, TCI America™
$33.56 - $95.06
Chemical Identifiers
| CAS | 112883-40-6 |
|---|---|
| Molecular Formula | C20H21NO4S |
| Molecular Weight (g/mol) | 371.45 |
| MDL Number | MFCD00062958 MFCD00037134 MFCD00062958 |
| InChI Key | BUBGAUHBELNDEW-GOSISDBHSA-N |
| Synonym | fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine |
| PubChem CID | 6992521 |
| IUPAC Name | (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid |
| SMILES | CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Chemical Identifiers
| 112883-40-6 | |
| 371.45 | |
| BUBGAUHBELNDEW-GOSISDBHSA-N | |
| 6992521 | |
| CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C20H21NO4S | |
| MFCD00062958 MFCD00037134 MFCD00062958 | |
| fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine | |
| (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid |
Specifications
| 112883-40-6 | |
| White-Yellow | |
| MFCD00062958 MFCD00037134 MFCD00062958 | |
| fmoc-d-met-oh, fmoc-d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl-d-methionine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylthio butanoic acid, d-methionine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylsulfanyl butanoic acid, n-fmoc-d-methionine, ambotzfaa1145, n-9-fluorenylmethyloxycarbonyl-d-methionine | |
| CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 371.45 | |
| 371.45 | |
| Crystalline Powder |
| 132°C | |
| C20H21NO4S | |
| 1 g | |
| BUBGAUHBELNDEW-GOSISDBHSA-N | |
| (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid | |
| 6992521 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine |
Safety and Handling
TSCA : No