missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-leucine 98.0+%, TCI America™
$29.83 - $82.02
Chemical Identifiers
| CAS | 114360-54-2 |
|---|---|
| Molecular Formula | C21H23NO4 |
| Molecular Weight (g/mol) | 353.42 |
| MDL Number | MFCD00062957 |
| InChI Key | CBPJQFCAFFNICX-TWYLJJHKNA-N |
| Synonym | fmoc-d-leu-oh, fmoc-d-leucine, n-fmoc-d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-d-leucine, n-9-fluorenylmethyloxycarbonyl-d-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-leucine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methylpentanoic acid |
| PubChem CID | 1549477 |
| IUPAC Name | (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid |
| SMILES | CC(C)C[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
Chemical Identifiers
| 114360-54-2 | |
| 353.42 | |
| CBPJQFCAFFNICX-TWYLJJHKNA-N | |
| 1549477 | |
| CC(C)C[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
| C21H23NO4 | |
| MFCD00062957 | |
| fmoc-d-leu-oh, fmoc-d-leucine, n-fmoc-d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-d-leucine, n-9-fluorenylmethyloxycarbonyl-d-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-leucine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methylpentanoic acid | |
| (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid |
Specifications
| 114360-54-2 | |
| White-Yellow | |
| MFCD00062957 | |
| fmoc-d-leu-oh, fmoc-d-leucine, n-fmoc-d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-d-leucine, n-9-fluorenylmethyloxycarbonyl-d-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-leucine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methylpentanoic acid | |
| CC(C)C[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O | |
| 353.42 | |
| 353.42 | |
| Crystalline Powder |
| 155°C | |
| C21H23NO4 | |
| 1 g | |
| CBPJQFCAFFNICX-TWYLJJHKNA-N | |
| (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid | |
| 1549477 | |
| ≥98.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-leucine |
Safety and Handling
TSCA : No