missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-leucine 98.0+%, TCI America™

Catalog No. F06031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06031G 1 g
F06035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06031G Supplier TCI America Supplier No. F06031G

Chemical Identifiers

CAS 114360-54-2
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00062957
InChI Key CBPJQFCAFFNICX-TWYLJJHKNA-N
Synonym fmoc-d-leu-oh, fmoc-d-leucine, n-fmoc-d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-d-leucine, n-9-fluorenylmethyloxycarbonyl-d-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-leucine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methylpentanoic acid
PubChem CID 1549477
IUPAC Name (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 1 g
Formula Weight 353.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-leucine