missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid 97.0+%, TCI America™
$96.94 - $294.52
Chemical Identifiers
| CAS | 104091-09-0 |
|---|---|
| Molecular Formula | C20H19NO6 |
| Molecular Weight (g/mol) | 369.373 |
| InChI Key | QEPWHIXHJNNGLU-QGZVFWFLSA-N |
| Synonym | N-Fmoc-D-glutamic Acid, Fmoc-D-Glu-OH |
| PubChem CID | 40501223 |
| IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O |
Chemical Identifiers
| 104091-09-0 | |
| 369.373 | |
| N-Fmoc-D-glutamic Acid, Fmoc-D-Glu-OH | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid |
| C20H19NO6 | |
| QEPWHIXHJNNGLU-QGZVFWFLSA-N | |
| 40501223 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O |
Specifications
| 104091-09-0 | |
| C20H19NO6 | |
| N-Fmoc-D-glutamic Acid, Fmoc-D-Glu-OH | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCC(=O)O)C(=O)O | |
| 369.373 | |
| 369.37 | |
| Crystalline Powder |
| White-Yellow | |
| 5 g | |
| QEPWHIXHJNNGLU-QGZVFWFLSA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanedioic acid | |
| 40501223 | |
| ≥97.0% (HPLC,T) | |
| N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-glutamic Acid |
Safety and Handling
TSCA : No