missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine 98.0+%, TCI America™

Catalog No. p-7129689
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04431G 1 g
F04435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04431G Supplier TCI America Supplier No. F04431G

Chemical Identifiers

CAS 95753-55-2
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD00057810
InChI Key RZRRJPNDKJOLHI-QFIPXVFZSA-N
Synonym fmoc-4-nitro-l-phenylalanine, fmoc-phe 4-no2-oh, fmoc-l-4-no2-phe-oh, fmoc-p-nitro-phe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-nitrophenyl propanoic acid, n-fmoc-4-nitro-l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-nitro-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-nitrophenyl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-3-4-nitrophenyl propanoic acid, fmoc-phe no2-oh
PubChem CID 7016054
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-nitrophenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(C=C1)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color Yellow
Quantity 1 g
Formula Weight 432.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine