missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine 95.0+%, TCI America™

Catalog No. p-7129967
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09021G 1 g
F09025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09021G Supplier TCI America Supplier No. F09021G
Only null left

Chemical Identifiers

CAS 169243-86-1
Molecular Formula C24H20FNO4
Molecular Weight (g/mol) 405.43
MDL Number MFCD00191197
InChI Key IXUMACXMEZBPJG-QFIPXVFZSA-N
Synonym fmoc-4-fluoro-l-phenylalanine, fmoc-l-4-fluorophenylalanine, fmoc-phe 4-f-oh, fmoc-l-4-fluorophe, fmoc-4-fluoro-l-phe, fmoc-4-fluoro-l-phe-oh, fmoc-l-phe 4-f-oh, fmoc-p-fluoro-phe-oh, l-4-fluoro-phenyl-n-fmoc-alanine, s-n-fmoc-4-fluorophenylalanine
PubChem CID 2734466
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-fluorophenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(F)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 184°C
Color White
Quantity 1 g
Formula Weight 405.43
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine
TSCA No
Recommended Storage Refrigerator