missing translation for 'onlineSavingsMsg'
Learn More

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-D-alanine 98.0+%, TCI America™

Catalog No. p-7129981
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
F0934200MG 200 mg
F09341G 1 g
2 options

Catalog No. F0934200MG

Supplier: TCI America F0934200MG

Only null left

Chemical Identifiers

CAS 220497-85-8
Molecular Formula C22H19NO5
Molecular Weight (g/mol) 377.40
MDL Number MFCD01311747
InChI Key AJXDCHXGNUFBRC-UHFFFAOYSA-N
Synonym fmoc-d-2-furylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-beta-2-furyl-d-ala-oh, n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid, fmoc-3-2-furyl-d-alanine, n-fmoc-3-2-furyl-d-alanine, fmoc-beta-2-furyl-d-ala-oh hplc
PubChem CID 16211163
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(furan-2-yl)propanoic acid
SMILES OC(=O)C(CC1=CC=CO1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 200 mg
Formula Weight 377.40
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-(2-furyl)-D-alanine