missing translation for 'onlineSavingsMsg'
Learn More

N-(9-Dimethylamino-1-oxononyl)-N-hydroxy-beta-alanine Methyl Ester 97.0+%, TCI America™

Catalog No. D40785MG
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40785MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40785MG Supplier TCI America Supplier No. D40785MG
Only null left

Chemical Identifiers

CAS 1239468-48-4
Molecular Formula C15H30N2O4
Molecular Weight (g/mol) 302.42
MDL Number MFCD22375684
InChI Key KDYRPQNFCURCQB-UHFFFAOYSA-N
Synonym NCDM-32b
PubChem CID 46901417
IUPAC Name methyl 3-[9-(dimethylamino)-N-hydroxynonanamido]propanoate
SMILES COC(=O)CCN(O)C(=O)CCCCCCCCN(C)C

Specifications

Color White-Yellow
Quantity 5 mg
Formula Weight 302.42
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(9-Dimethylamino-1-oxononyl)-N-hydroxy-beta-alanine Methyl Ester
TSCA No
Recommended Storage Refrigerator