missing translation for 'onlineSavingsMsg'
Learn More

N-(9-Cyclopropyl-1-oxononyl)-N-hydroxy-beta-alanine 80.0+%, TCI America™

Catalog No. c31345mg
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31345MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31345MG Supplier TCI America Supplier No. C31345MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1453071-47-0
Molecular Formula C15H27NO4
Molecular Weight (g/mol) 285.384
InChI Key NEHSERYKENINRH-UHFFFAOYSA-N
Synonym NCDM-64
PubChem CID 71735843
IUPAC Name 3-[9-cyclopropylnonanoyl(hydroxy)amino]propanoic acid
SMILES C1CC1CCCCCCCCC(=O)N(CCC(=O)O)O

Specifications

Melting Point 87°C
Color White
Quantity 5 mg
Formula Weight 285.38
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(9-Cyclopropyl-1-oxononyl)-N-hydroxy-beta-alanine
TSCA No
Recommended Storage Freezer