missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Bromohexyl)phthalimide 98.0+%, TCI America™

Catalog No. p-7123886
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49875G 5 g
B498725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49875G Supplier TCI America Supplier No. B49875G
Only null left

Chemical Identifiers

CAS 24566-79-8
Molecular Formula C14H16BrNO2
Molecular Weight (g/mol) 310.191
MDL Number MFCD00023098
InChI Key OAZFTIPKNPTDIO-UHFFFAOYSA-N
PubChem CID 141120
IUPAC Name 2-(6-bromohexyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr

Specifications

Melting Point 58°C
Color White
Boiling Point 188°C
Quantity 5 g
Synonym n-6-bromohexyl phthalimide, 2-6-bromohexyl isoindoline-1,3-dione, n-6-bromohexyl-phthalimide, 2-6-bromohexyl-1h-isoindole-1,3 2h-dione, 2-6-bromohexyl isoindole-1,3-dione, 2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-6-bromo-hexyl-isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-6-bromohexyl, 1h-isoindole-1,3 2h-dione, 2-6-bromohexyl, 6-n-phthalimido hexyl bromide
Formula Weight 310.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(6-Bromohexyl)phthalimide