missing translation for 'onlineSavingsMsg'
Learn More

N,5-Dimethyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1031843-22-7
Molecular Formula C12H15N3
Molecular Weight (g/mol) 201.27
MDL Number MFCD11109316
InChI Key PYMKNYBFTGMBJN-UHFFFAOYSA-N
Synonym methyl 5-methyl-1-phenylpyrazol-4-yl methyl amine, 4-methyl aminomethyl-5-methyl-1-phenyl-1h-pyrazole, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methylamine, methyl 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, n,5-dimethyl-1-phenyl-1h-pyrazol-4-ylmethyl amine, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine
PubChem CID 33589450
IUPAC Name N-methyl-1-(5-methyl-1-phenylpyrazol-4-yl)methanamine
SMILES CNCC1=C(C)N(N=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC15546CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC15546DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1031843-22-7
201.27
PYMKNYBFTGMBJN-UHFFFAOYSA-N
33589450
CNCC1=C(C)N(N=C1)C1=CC=CC=C1
C12H15N3
MFCD11109316
methyl 5-methyl-1-phenylpyrazol-4-yl methyl amine, 4-methyl aminomethyl-5-methyl-1-phenyl-1h-pyrazole, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methylamine, methyl 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, n,5-dimethyl-1-phenyl-1h-pyrazol-4-ylmethyl amine, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine
N-methyl-1-(5-methyl-1-phenylpyrazol-4-yl)methanamine

Specifications

1031843-22-7
100.0
C12H15N3
methyl 5-methyl-1-phenylpyrazol-4-yl methyl amine, 4-methyl aminomethyl-5-methyl-1-phenyl-1h-pyrazole, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methylamine, methyl 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, n,5-dimethyl-1-phenyl-1h-pyrazol-4-ylmethyl amine, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine
CNCC1=C(C)N(N=C1)C1=CC=CC=C1
201.27
201.27
N, 5-Dimethyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine
96.5
97%
MFCD11109316
PYMKNYBFTGMBJN-UHFFFAOYSA-N
N-methyl-1-(5-methyl-1-phenylpyrazol-4-yl)methanamine
33589450
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents