missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Bromopentyl)phthalimide 98.0+%, TCI America™

Catalog No. p-7123885
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49865G 5 g
B498625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49865G Supplier TCI America Supplier No. B49865G

Chemical Identifiers

CAS 954-81-4
Molecular Formula C13H14BrNO2
Molecular Weight (g/mol) 296.164
MDL Number MFCD00060522
InChI Key QKVHAKICMNABGB-UHFFFAOYSA-N
Synonym n-5-bromopentyl phthalimide, 2-5-bromopentyl isoindole-1,3-dione, n-5-bromopentyl-phthalimide, 2-5-bromopentyl isoindoline-1,3-dione, 2-5-bromo-pentyl-isoindole-1,3-dione, 2-5-bromopentyl-1h-isoindole-1,3 2h-dione, 2-5-bromopentyl-2,3-dihydro-1h-isoindole-1,3-dione, 5-phthalimidopentyl bromide, 1-bromo-5-phthalimidopentane
PubChem CID 136770
IUPAC Name 2-(5-bromopentyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCBr

Specifications

Melting Point 62°C
Quantity 5 g
Formula Weight 296.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(5-Bromopentyl)phthalimide