missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Pentynyl)phthalimide, 97%

Catalog No. AC425730050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425730050 5 g Glass bottle
AC425730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425730050 Supplier Thermo Scientific Chemicals Supplier No. 425730050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6097-07-0 | C13H11NO2 | 213.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6097-07-0
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.23
MDL Number MFCD06798110
InChI Key YNZIPXLLPFYDGM-UHFFFAOYSA-N
Synonym n-4-pentynyl phthalimide, 2-pent-4-ynyl isoindoline-1,3-dione, 1-phthalimido-4-pentyne, 2-pent-4-yn-1-yl isoindoline-1,3-dione, n-4-pentynyl phthalimide 97, 2-pent-4-yn-1-yl isoindole-1,3-dione, 5-phthalimido-1-pentyne, 2-pent-4-ynyl-isoindole-1,3-dione
PubChem CID 11769822
IUPAC Name 2-pent-4-ynylisoindole-1,3-dione
SMILES C#CCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 87.0°C to 91.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 213.23
Percent Purity 97%
Chemical Name or Material N-(4-Pentynyl)phthalimide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only