missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Oxocyclohexyl)phthalimide 98.0+%, TCI America™
Supplier: TCI America O036525G
Specifications
| N-(4-Oxocyclohexyl)phthalimide | |
| 144°C | |
| C14H13NO3 | |
| 25 g | |
| PWUJQPNLEZZILN-UHFFFAOYSA-N | |
| 2-(4-oxocyclohexyl)isoindole-1,3-dione | |
| 11998391 | |
| ≥98.0% (GC,N) |
| 104618-32-8 | |
| White | |
| MFCD00158659 | |
| 4-Phthalimidocyclohexanone | |
| C1CC(=O)CCC1N2C(=O)C3=CC=CC=C3C2=O | |
| 243.262 | |
| 243.26 | |
| Crystalline Powder |
Chemical Identifiers
| 104618-32-8 | |
| 243.262 | |
| PWUJQPNLEZZILN-UHFFFAOYSA-N | |
| 11998391 | |
| C1CC(=O)CCC1N2C(=O)C3=CC=CC=C3C2=O |
| C14H13NO3 | |
| MFCD00158659 | |
| 4-Phthalimidocyclohexanone | |
| 2-(4-oxocyclohexyl)isoindole-1,3-dione |
Safety and Handling
TSCA : No