missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Nitrophenyl)-3-oxobutyramide 98.0+%, TCI America™

Catalog No. N0990200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0990200MG 200 mg
N09901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0990200MG Supplier TCI America Supplier No. N0990200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4835-39-6
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.2
MDL Number MFCD00452522
InChI Key KCXJQNDNGLRYBN-UHFFFAOYSA-N
Synonym 4′C-Nitroacetoacetanilide
PubChem CID 78547
IUPAC Name N-(4-nitrophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 122°C
Color Yellow
Quantity 200 mg
Formula Weight 222.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Nitrophenyl)-3-oxobutyramide
EINECSNumber (3)-0393
TSCA No