missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Methoxybenzylidene)-4-acetoxyaniline 99.0+%, TCI America™

Catalog No. m058125g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M058125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M058125G Supplier TCI America Supplier No. M058125G
Only null left

Chemical Identifiers

CAS 10484-13-6
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.30
MDL Number MFCD00041911
InChI Key HOYWVKUPOCFKOT-UHFFFAOYSA-N
Synonym apapa, n-4-methoxybenzylidene-4-acetoxyaniline, 4'-methoxybenzylidene-4-acetoxyaniline, p-anisylidene-p-aminophenyl acetate, p-n-anisylideneamino phenyl acetate, 4-methoxybenzylidene-4'-aminophenyl acetate, p-p-methoxybenzylidene amino phenyl acetate, n-p-methoxybenzylidene-p-aminophenyl acetate, anisylidene p-aminophenylacetate, p-anisylideneamino phenyl acetate
PubChem CID 97495
IUPAC Name 4-{[(4-methoxyphenyl)methylidene]amino}phenyl acetate
SMILES COC1=CC=C(C=NC2=CC=C(OC(C)=O)C=C2)C=C1

Specifications

Quantity 25 g
Formula Weight 269.30
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Methoxybenzylidene)-4-acetoxyaniline