missing translation for 'onlineSavingsMsg'
Learn More

N-(4'-Iodobiphenyl-4-yl)-N-(m-tolyl)aniline 98.0+%, TCI America™

Catalog No. I04901G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04901G Supplier TCI America Supplier No. I04901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 195443-34-6
Molecular Formula C25H20IN
Molecular Weight (g/mol) 461.346
MDL Number MFCD00671551
InChI Key AKQZSFMSAULQQN-UHFFFAOYSA-N
Synonym N-(4′C-Iodobiphenyl-4-yl)-N-phenyl-N-(m-tolyl)amine, 4′C-Iodo-4-[N-phenyl-N-(m-tolyl)amino]biphenyl
PubChem CID 10718819
IUPAC Name N-[4-(4-iodophenyl)phenyl]-3-methyl-N-phenylaniline
SMILES CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C4=CC=C(C=C4)I

Specifications

Melting Point 160°C
Quantity 1 g
Formula Weight 461.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4′-Iodobiphenyl-4-yl)-N-(m-tolyl)aniline